Please login to your online account to display your discounted pricing

2,5-Dimethyl-3-hexyne-2,5-diol, 98%, ACROS Organics™

$75.72 - $75.72

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethyl-3-hexyne-2, 5-diol
CAS 142-30-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H14O2
Linear Formula HOC(CH3)2C2CC(CH3)2OH
Formula Weight 142.2
MDL Number MFCD00004468
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC207025000

  SDS
thermo fisher scientific
AC207025000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC207021000

  SDS
thermo fisher scientific
207021000
Plastic bottle 100g Each for $75.72
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 142.2
Melting Point >89°C
Physical Form Crystalline Flakes or Powder
Quantity 500g
Color White to light yellow
Beilstein 01, 501
CAS Max % 100.0
Linear Formula HOC(CH3)2C2CC(CH3)2OH
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 142-30-3
Molecular Formula C8H14O2
CAS Min % 97.5
MDL Number MFCD00004468
Packaging Plastic bottle
Chemical Name or Material 2, 5-Dimethyl-3-hexyne-2, 5-diol