Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-3-hexanol, 98%

$33.76 - $33.76

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-3-hexanol
CAS 19550-07-3
Molecular Formula C8H18O
Formula Weight 130.23
MDL Number MFCD00026452
Synonym Isobutyl isopropyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2057803

  SDS
alfa aesar™
B2057803
1g Each for $33.76
Description & Specifications

Specifications

Formula Weight 130.23
Molecular Formula C8H18O
Chemical Name or Material 2,5-Dimethyl-3-hexanol
Quantity 1g
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Isobutyl isopropyl carbinol
CAS 19550-07-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
MDL Number MFCD00026452
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.