missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,5-Dimethyl-3-furyl)methylamine, 97%, Maybridge™

Chemical Identifiers

CAS 306934-85-0
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD02677701
InChI Key AGQXLVABIKZJJG-UHFFFAOYSA-N
Synonym 1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethylfuran-3-yl methanamine, 2,5-dimethylfuran-3-yl methyl amine, 3-aminomethyl-2,5-dimethylfuran, 3-furanmethanamine,2,5-dimethyl, sz9
PubChem CID 2776162
IUPAC Name (2,5-dimethylfuran-3-yl)methanamine
SMILES CC1=CC(=C(O1)C)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC01113CB
SDS View Documents
Fisher Scientific Company
CC01113CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01113DA
SDS View Documents
Fisher Scientific Company
CC01113DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

306934-85-0
125.171
AGQXLVABIKZJJG-UHFFFAOYSA-N
2776162
CC1=CC(=C(O1)C)CN
C7H11NO
MFCD02677701
1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethylfuran-3-yl methanamine, 2,5-dimethylfuran-3-yl methyl amine, 3-aminomethyl-2,5-dimethylfuran, 3-furanmethanamine,2,5-dimethyl, sz9
(2,5-dimethylfuran-3-yl)methanamine

Specifications

Amber Glass Bottle
306934-85-0
100.0
C7H11NO
1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethylfuran-3-yl methanamine, 2,5-dimethylfuran-3-yl methyl amine, 3-aminomethyl-2,5-dimethylfuran, 3-furanmethanamine,2,5-dimethyl, sz9
CC1=CC(=C(O1)C)CN
125.171
125.17
(2, 5-Dimethyl-3-furyl)methylamine
250mg
89.5
90%
MFCD02677701
AGQXLVABIKZJJG-UHFFFAOYSA-N
(2,5-dimethylfuran-3-yl)methanamine
2776162
90%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS