missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,5-Dimethyl-3-furyl)methylamine, 97%, Maybridge

Manufacturer:  Thermo Scientific CC01113CB

 View more versions of this product

Catalog No. CC01113CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(2, 5-Dimethyl-3-furyl)methylamine
250mg
89.5
90%
MFCD02677701
AGQXLVABIKZJJG-UHFFFAOYSA-N
(2,5-dimethylfuran-3-yl)methanamine
2776162
90%
Amber Glass Bottle
306934-85-0
100.0
C7H11NO
2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfuran-3-yl methanamine, 3-furanmethanamine,2,5-dimethyl, 3-aminomethyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfuran-3-yl methyl amine, sz9
CC1=CC(=C(O1)C)CN
125.171
125.17

Chemical Identifiers

306934-85-0
125.171
AGQXLVABIKZJJG-UHFFFAOYSA-N
2776162
CC1=CC(=C(O1)C)CN
C7H11NO
MFCD02677701
2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfuran-3-yl methanamine, 3-furanmethanamine,2,5-dimethyl, 3-aminomethyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfuran-3-yl methyl amine, sz9
(2,5-dimethylfuran-3-yl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents