Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-3-furoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-3-furoyl chloride
CAS 50990-93-7
Molecular Formula C7H7ClO2
Formula Weight 158.59
MDL Number MFCD00221073
UN Number UN3265
Beilstein 116198
Synonym 2,4-Dimethylfuran-3-carbonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1842203

  SDS
alfa aesar™
L1842203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1842206

  SDS
alfa aesar™
L1842206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.59
Chemical Name or Material 2,5-Dimethyl-3-furoyl chloride
MDL Number MFCD00221073
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Boiling Point 87° to 89°C (15mmHg)
CAS 50990-93-7
UN Number UN3265
Beilstein 116198
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Molecular Formula C7H7ClO2
Synonym 2,4-Dimethylfuran-3-carbonyl chloride
Melting Point 20°C

Applications
They are admirably suited for studies on orientation of the furan nucleus, both by absolute and relative methods. In addition they make beta-substituted furans more accessible.

Solubility
Reacts with water. Soluble in chloroform and ethyl acetate.

Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.