missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-3-furansulfonyl chloride, 97%, Maybridge™

Chemical Identifiers

CAS 166964-26-7
Molecular Formula C6H7ClO3S
Molecular Weight (g/mol) 194.63
MDL Number MFCD04115370
InChI Key PIXAQZAVANAGLE-UHFFFAOYSA-N
Synonym 2,5-dimethyl 3-furyl chlorosulfone, 2,5-dimethyl-3-furansulfonyl chloride, 2,5-dimethyl-furan-3-sulfonylchloride, 2,5-dimethylfuran 3-sulfonyl chloride, 3-furansulfonylchloride, 2,5-dimethyl
PubChem CID 2795024
IUPAC Name 2,5-dimethylfuran-3-sulfonyl chloride
SMILES CC1=CC(=C(C)O1)S(Cl)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC01103CB
SDS View Documents
Fisher Scientific Company
CC01103CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01103DA
SDS View Documents
Fisher Scientific Company
CC01103DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

166964-26-7
194.63
PIXAQZAVANAGLE-UHFFFAOYSA-N
2795024
CC1=CC(=C(C)O1)S(Cl)(=O)=O
C6H7ClO3S
MFCD04115370
2,5-dimethyl 3-furyl chlorosulfone, 2,5-dimethyl-3-furansulfonyl chloride, 2,5-dimethyl-furan-3-sulfonylchloride, 2,5-dimethylfuran 3-sulfonyl chloride, 3-furansulfonylchloride, 2,5-dimethyl
2,5-dimethylfuran-3-sulfonyl chloride

Specifications

Amber Glass Bottle
166964-26-7
100.0
C6H7ClO3S
2,5-dimethyl 3-furyl chlorosulfone, 2,5-dimethyl-3-furansulfonyl chloride, 2,5-dimethyl-furan-3-sulfonylchloride, 2,5-dimethylfuran 3-sulfonyl chloride, 3-furansulfonylchloride, 2,5-dimethyl
CC1=CC(=C(C)O1)S(Cl)(=O)=O
194.63
194.64
2, 5-Dimethyl-3-furansulfonyl chloride
250mg
96.5
97%
MFCD04115370
PIXAQZAVANAGLE-UHFFFAOYSA-N
2,5-dimethylfuran-3-sulfonyl chloride
2795024
97%
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Danger

SDS
Product Certifications