Please login to your online account to display your discounted pricing

2,5-Dimethyl-3-furansulfonyl chloride, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC01103DA

 View more versions of this product

Catalog No. CC01103DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.64
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H7ClO3S
CAS 166964-26-7
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Chemical Name or Material 2, 5-Dimethyl-3-furansulfonyl chloride
Packaging Amber Glass Bottle
MDL Number MFCD04115370
CAS Min % 96.5
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Specifications

Chemical Name or Material 2, 5-Dimethyl-3-furansulfonyl chloride
CAS 166964-26-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7ClO3S
Formula Weight 194.64
MDL Number MFCD04115370
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage.