Please login to your online account to display your discounted pricing

2,5-Dimethyl-3,4-diphenylcyclopentadienone dimer, 97%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  33496-0250

 View more versions of this product

Catalog No. AC334960250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Fine Powder
Formula Weight 520.67
Quantity 25g
Color White
TSCA TSCA
Melting Point 185° to 190°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 2, 5-Dimethyl-3, 4-diphenylcyclopentadienone dimer
Name Note 97%
Assay Percent Range 97%
Molecular Formula C38H32O2
MDL Number MFCD00001408
CAS 38883-84-0

Specifications

Chemical Name or Material 2, 5-Dimethyl-3, 4-diphenylcyclopentadienone dimer
Name Note 97%
CAS 38883-84-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C38H32O2
Formula Weight 520.67
MDL Number MFCD00001408