Please login to your online account to display your discounted pricing

2,5-Dimethyl-3,4-diphenylcyclopentadienone dimer, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethyl-3, 4-diphenylcyclopentadienone dimer
Name Note 97%
CAS 38883-84-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C38H32O2
Formula Weight 520.67
MDL Number MFCD00001408
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334960250

  SDS
thermo fisher scientific
33496-0250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fine Powder
Formula Weight 520.67
Quantity 25g
Color White
TSCA TSCA
Melting Point 185° to 190°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 2, 5-Dimethyl-3, 4-diphenylcyclopentadienone dimer
Name Note 97%
Assay Percent Range 97%
Molecular Formula C38H32O2
MDL Number MFCD00001408
CAS 38883-84-0