Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-3(2H)-furanone, 94%

$41.55 - $574.15

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-3 (2H)-furanone
CAS 14400-67-0
Molecular Formula C6H8O2
Formula Weight 112.13
MDL Number MFCD00052571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1218203

  SDS
alfa aesar™
A1218203
1g Each for $41.55

AAA1218206

  SDS
alfa aesar™
A1218206
5g Each for $155.47

AAA1218214

  SDS
alfa aesar™
A1218214
25g Each for $574.15
Description & Specifications

Specifications

Formula Weight 112.13
Chemical Name or Material 2,5-Dimethyl-3 (2H)-furanone
TSCA TSCA
Assay 94%
Molecular Formula C6H8O2
Boiling Point 259° to 261°C
MDL Number MFCD00052571
CAS 14400-67-0
Quantity 1g
Flash Point 68°C (154°F)
Odor Sweet
Health Hazard 1 Warning
Safety and Handling

Warning