Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-2-hexanol, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-2-hexanol
CAS 3730-60-7
Molecular Formula C8H18O
Formula Weight 130.23
MDL Number MFCD00021814
UN Number UN1987
Beilstein 1732761
Synonym Dimethyl isoamyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2035206

  SDS
alfa aesar™
B2035206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2035214

  SDS
alfa aesar™
B2035214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2035222

  SDS
alfa aesar™
B2035222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.23
Beilstein 1732761
MDL Number MFCD00021814
Synonym Dimethyl isoamyl carbinol
Assay 98%
CAS 3730-60-7
Flash Point 52°C (125°F)
Molecular Formula C8H18O
Quantity 5g
UN Number UN1987
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Chemical Name or Material 2,5-Dimethyl-2-hexanol
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Boiling Point 155° to 156°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.