missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 349900050

 View more versions of this product

Catalog No. AC349900050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
Specifications

Specifications

2, 5-Dimethyl-2, 5-di(tert-butylperoxy)hexane
6.0°C
55.0°C to 57.0°C (7.0 mmHg)
90% min. (GC)
C16H34O4
5 g
varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50
DMWVYCCGCQPJEA-UHFFFAOYSA-N
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
6545
290.44
78-63-7
0.8700g/mL
68°C
Plastic bottle
1.4200 to 1.4240
0.87
Solubility in water: immiscible. Other solubilities: soluble in organic solvents,soluble in alcohol
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
290.44
6.4 mPa.s (20°C)
92%

Chemical Identifiers

78-63-7
290.44
varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
C16H34O4
DMWVYCCGCQPJEA-UHFFFAOYSA-N
6545
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Heating may cause a fire.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep/Store away from clothing/ combustible materials.
Store in a well-v

GHS Signal Word: Danger

EINECSNumber : 201-128-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only