Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-1-phenylpyrrole, 98+%

$42.58 - $42.58

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-1-phenylpyrrole
CAS 83-24-9
Molecular Formula C12H13N
Formula Weight 171.24g/mol
MDL Number MFCD00022464
Beilstein 124370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1461406

 
alfa aesar™
A14614-06
5g Each for $42.58
Description & Specifications

Specifications

Melting Point 48° to 52°C
Boiling Point 155° to 160°C (15mmHg)
Quantity 5g
Chemical Name or Material 2,5-Dimethyl-1-phenylpyrrole
CAS 83-24-9
Assay ≥98%
Molecular Formula C12H13N
Formula Weight 171.24g/mol
MDL Number MFCD00022464
Beilstein 124370
Recommended Storage 2° to 8°C
TSCA TSCA
Safety and Handling

2° to 8°C