Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole, 97%

$67.66 - $241.08

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole
CAS 5044-22-4
Molecular Formula C12H12N2O2
Formula Weight 216.24
MDL Number MFCD00085038
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2514503

  SDS
alfa aesar™
B2514503
1g Each for $67.66

AAB2514506

  SDS
alfa aesar™
B2514506
5g Each for $241.08
Description & Specifications

Specifications

Formula Weight 216.24
Chemical Name or Material 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole
Assay 97%
Molecular Formula C12H12N2O2
Melting Point 145° to 148°C
MDL Number MFCD00085038
CAS 5044-22-4
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.