Please login to your online account to display your discounted pricing

2,5-Dimethyl-1,3-oxazole-4-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC26801DA

 View more versions of this product

Catalog No. CC26801DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 141.13
Quantity 1g
Assay 97%
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Molecular Formula C6H7NO3
Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carboxylic acid
CAS 23000-14-8
CAS Min % 96.5
MDL Number MFCD03011595
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carboxylic acid
CAS 23000-14-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7NO3
Formula Weight 141.13
MDL Number MFCD03011595
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents