missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-1,3-oxazole-4-carbonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC26802DA

 View more versions of this product

Catalog No. CC26802DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 5-Dimethyl-1, 3-oxazole-4-carbonyl chloride
96.5
97%
C6H6ClNO2
1g
XZXCVPKMCPXVFF-UHFFFAOYSA-N
dimethyl-1,3-oxazole-4-carbonyl chloride
7018020
97%
197719-27-0
100.0
Amber Glass Bottle
MFCD04972642
2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
CC1=NC(C(Cl)=O)=C(C)O1
159.57
159.57

Chemical Identifiers

197719-27-0
159.57
XZXCVPKMCPXVFF-UHFFFAOYSA-N
7018020
CC1=NC(C(Cl)=O)=C(C)O1
C6H6ClNO2
MFCD04972642
2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
dimethyl-1,3-oxazole-4-carbonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions