Please login to your online account to display your discounted pricing

(2,5-Dimethyl-1,3-oxazol-4-yl)methylamine, ≥97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC26813CB

 View more versions of this product

Catalog No. CC26813CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.16
Quantity 250mg
Assay 97%
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C6H10N2O
CAS 859850-62-7
Packaging Amber Glass Bottle
Chemical Name or Material (2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine
MDL Number MFCD08271883
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine
CAS 859850-62-7
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H10N2O
Formula Weight 126.16
MDL Number MFCD08271883
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents