missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,5-Dimethyl-1,3-oxazol-4-yl)methylamine, ≥97%, Maybridge

$590.00 - $590.00

Chemical Identifiers

CAS 859850-62-7
Assay Percent Range 97%
Molecular Formula C6H10N2O
Formula Weight 126.16
MDL Number MFCD08271883
Chemical Name or Material (2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC26813CB SDS Fisher Scientific Company
CC26813CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC26813DA SDS Fisher Scientific Company
CC26813DA
1g Each for $590.00
Specifications Description & Specifications

Chemical Identifiers

859850-62-7
C6H10N2O
MFCD08271883
97%
97%
126.16
(2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine

Specifications

126.16
97%
859850-62-7
MFCD08271883
97%
250mg
C6H10N2O
Amber Glass Bottle
(2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.