missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,5-Dimethyl-1,3-oxazol-4-yl)methanol, ≥97%, Maybridge

Chemical Identifiers

CAS 92901-94-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H9NO2
Formula Weight 127.14
MDL Number MFCD07772804
Chemical Name or Material (2, 5-Dimethyl-1, 3-oxazol-4-yl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC26809CB SDS Fisher Scientific Company
CC26809CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC26809DA SDS Fisher Scientific Company
CC26809DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

92901-94-5
100.0
C6H9NO2
MFCD07772804
97%
96.5
97%
127.14
(2, 5-Dimethyl-1, 3-oxazol-4-yl)methanol

Specifications

127.14
100.0
MFCD07772804
96.5
C6H9NO2
97%
250mg
97%
92901-94-5
Amber Glass Bottle
(2, 5-Dimethyl-1, 3-oxazol-4-yl)methanol

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning