Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-1,3,4-thiadiazole, 99+%

$41.55 - $140.08

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-1,3,4-thiadiazole
CAS 27464-82-0
Molecular Formula C4H6N2S
Formula Weight 114.17
MDL Number MFCD00020722
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1722706

 
alfa aesar™
A1722706
5g Each for $41.55

AAA1722714

 
alfa aesar™
A1722714
25g Each for $140.08
Description & Specifications

Specifications

Formula Weight 114.17
Chemical Name or Material 2,5-Dimethyl-1,3,4-thiadiazole
TSCA TSCA
Assay ≥99%
Molecular Formula C4H6N2S
Melting Point 61° to 65°C
MDL Number MFCD00020722
CAS 27464-82-0
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.