Please login to your online account to display your discounted pricing

2,5-Dimethoxytoluene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethoxytoluene
CAS 24599-58-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O2
Linear Formula CH3C6H3(OCH3)2
Formula Weight 152.19
MDL Number MFCD00041901
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC407930250

  SDS
thermo fisher scientific
AC407930250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC407931000

  SDS
thermo fisher scientific
407931000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 152.19
Quantity 25g
Color Clear light yellow
Density 1.0400g/mL
CAS Max % 100.0
Chemical Name or Material 2, 5-Dimethoxytoluene
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C9H12O2
Linear Formula CH3C6H3(OCH3)2
MDL Number MFCD00041901
CAS 24599-58-4
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.