Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxytoluene, 98+%

$197.03 - $197.03

Chemical Identifiers

Chemical Name or Material 2,5-Dimethoxytoluene
CAS 24599-58-4
Molecular Formula C9H12O2
Formula Weight 152.19
MDL Number MFCD00041901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2163114

 
alfa aesar™
B2163114
25g Each for $197.03
Description & Specifications

Specifications

Formula Weight 152.19
Chemical Name or Material 2,5-Dimethoxytoluene
Assay ≥98%
MDL Number MFCD00041901
Health Hazard 2 GHS H Statement
H315
Causes skin irritation.
Flash Point 102°C (216°F)
Odor Aromatic
Melting Point 19° to 21°C
TSCA TSCA
Molecular Formula C9H12O2
CAS 24599-58-4
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P264-P362-P321-P332+P313-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Boiling Point 220°C
Quantity 25g

Applications
2,5-Dimethoxytoluene may be used as a reagent in the synthesis of 2,5-dimethoxy-4-methylbenzaldehyde-7-14C. Its IR spectrum has been used to establish the methyl group migration in the acid catalyzed rearrangement of 4-methoxy-4-methylcyclohexadienone.

Solubility
Not miscible or difficult to mix with water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280-P264-P362-P321-P332+P313-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H315
Causes skin irritation.