Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxythiophenol, 95%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethoxythiophenol
CAS 1483-27-8
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00052738
UN Number UN3334
Beilstein 1867842
Synonym 2,5-Dimethoxybenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1164106

  SDS
alfa aesar™
L1164106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material 2,5-Dimethoxythiophenol
MDL Number MFCD00052738
Assay 95%
Molecular Formula C8H10O2S
CAS 1483-27-8
UN Number UN3334
Beilstein 1867842
Synonym 2,5-Dimethoxybenzenethiol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
Flash Point 122°C (252°F)
Odor Stench
Boiling Point 278°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.