missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxythiophenol, 95%

Manufacturer:  Alfa Aesar™ L1164103

 View more versions of this product

Catalog No. AAL1164103



Specifications

Specifications

2,5-Dimethoxythiophenol
Stench
1g
C8H10O2S
UN3334
2,5-dimethoxythiophenol, 2,5-dimethoxybenzene-1-thiol, benzenethiol, 2,5-dimethoxy, pubchem6852, acmc-1cgdz, 3,6-dimethoxybenzenethiol, #, 2,5-dimethoxyphenyl mercaptan, 2,5-dimethoxythiophenol
COC1=CC(=C(C=C1)OC)S
170.226
170.23
122°C (252°F)
278°C
1483-27-8
MFCD00052738
1867842
SESUUAOAUZDHHP-UHFFFAOYSA-N
2,5-dimethoxybenzenethiol
519009
95%

Chemical Identifiers

1483-27-8
170.226
SESUUAOAUZDHHP-UHFFFAOYSA-N
519009
COC1=CC(=C(C=C1)OC)S
C8H10O2S
MFCD00052738
2,5-dimethoxythiophenol, 2,5-dimethoxybenzene-1-thiol, benzenethiol, 2,5-dimethoxy, pubchem6852, acmc-1cgdz, 3,6-dimethoxybenzenethiol, #, 2,5-dimethoxyphenyl mercaptan, 2,5-dimethoxythiophenol
2,5-dimethoxybenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.

Documents