Learn More
2,5-Dimethoxytetrahydrofuran, cis + trans, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1168722
Description
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
2,5-Dimethoxytetrahydrofuran | |
cis + trans | |
1.02 | |
100 g | |
C6H12O3 | |
UN3271 | |
2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf | |
GFISDBXSWQMOND-UHFFFAOYSA-N | |
2,5-dimethoxyoxolane | |
79098 | |
98% |
-45°C | |
696-59-3 | |
145°C to 147°C | |
1.418 | |
MFCD00005359 | |
104261 | |
Miscible with methanol,diethyl ether and tetrahydrofuran. Immiscible with water. | |
COC1CCC(O1)OC | |
132.159 | |
132.16 |
Chemical Identifiers
696-59-3 | |
132.159 | |
GFISDBXSWQMOND-UHFFFAOYSA-N | |
79098 | |
COC1CCC(O1)OC |
C6H12O3 | |
MFCD00005359 | |
2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf | |
2,5-dimethoxyoxolane |
Safety and Handling
GHS H Statement
H330-H226-H302-H315-H319-H335
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P370+P378q-P501c
H226-H319-H331
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 211-797-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only