missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115675000
Specifications
2, 5-Dimethoxytetrahydrofuran | |
35°C | |
Glass bottle | |
1.021 | |
Colorless to Yellow | |
500g | |
696-59-3 | |
100.0 | |
C6H12O3 | |
02,144 | |
Solubility in water: 350g/L (20°C). Other solubilities: miscible with alcohols and most common organic,solvents | |
COC1CCC(O1)OC | |
132.16 | |
132.16 | |
99% |
1.0210g/mL | |
Authentic | |
1.4160 to 1.419 | |
145°C to 148°C | |
-45°C | |
99% | |
98.5 | |
99% | |
MFCD00005359 | |
2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, 2,5-dimethoxytetrahydrofurane, dimethoxytetrahydrofuran van, dmthf, furan, tetrahydro-2,5-dimethoxy, furan,5-dimethoxy, protectol dmt, tetrahydro-2,5-dimethoxyfuran | |
GFISDBXSWQMOND-UHFFFAOYSA-N | |
2,5-dimethoxyoxolane | |
79098 | |
Liquid |
Chemical Identifiers
696-59-3 | |
132.16 | |
GFISDBXSWQMOND-UHFFFAOYSA-N | |
79098 | |
COC1CCC(O1)OC |
C6H12O3 | |
MFCD00005359 | |
2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, 2,5-dimethoxytetrahydrofurane, dimethoxytetrahydrofuran van, dmthf, furan, tetrahydro-2,5-dimethoxy, furan,5-dimethoxy, protectol dmt, tetrahydro-2,5-dimethoxyfuran | |
2,5-dimethoxyoxolane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Toxic if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INH
GHS Signal Word: Danger
EINECSNumber : 211-797-1
TSCA : TSCA