Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxyphenyl isocyanate, 99%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethoxyphenyl isocyanate
CAS 56309-62-7
Molecular Formula C9H9NO3
Formula Weight 179.18
MDL Number MFCD00002006
UN Number UN2811
Beilstein 3267303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1138103

 
alfa aesar™
L1138103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1138106

 
alfa aesar™
L1138106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1138114

 
alfa aesar™
L1138114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.18
Chemical Name or Material 2,5-Dimethoxyphenyl isocyanate
Assay 99%
MDL Number MFCD00002006
CAS 56309-62-7
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 38° to 42°C
Molecular Formula C9H9NO3
Beilstein 3267303
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 152° to 154°C (26mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C