missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxyphenyl isocyanate, 99%

Manufacturer:  Alfa Aesar™ L1138103

 View more versions of this product

Catalog No. AAL1138103



Specifications

Specifications

2,5-Dimethoxyphenyl isocyanate
152°C to 154°C (26mmHg)
1g
C9H9NO3
UN2811
2,5-dimethoxyphenyl isocyanate, 2,5-dimethoxyphenylisocyanate, acmc-1az6v, 2,5-dimethoxybenzenisocyanate, 2-isocyanato-1,4-dimethoxy-benzene, 2,5-dimethoxyphenyl isocyanate, benzene, 2-isocyanato-1,4-dimethoxy-9ci
COC1=CC(=C(C=C1)OC)N=C=O
179.175
179.18
>110°C (230°F)
38°C to 42°C
56309-62-7
MFCD00002006
3267303
XNQBFHMCUNRKPM-UHFFFAOYSA-N
2-isocyanato-1,4-dimethoxybenzene
4178425
99%

Chemical Identifiers

56309-62-7
179.175
XNQBFHMCUNRKPM-UHFFFAOYSA-N
4178425
COC1=CC(=C(C=C1)OC)N=C=O
C9H9NO3
MFCD00002006
2,5-dimethoxyphenyl isocyanate, 2,5-dimethoxyphenylisocyanate, acmc-1az6v, 2,5-dimethoxybenzenisocyanate, 2-isocyanato-1,4-dimethoxy-benzene, 2,5-dimethoxyphenyl isocyanate, benzene, 2-isocyanato-1,4-dimethoxy-9ci
2-isocyanato-1,4-dimethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents