Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxycinnamic acid, predominantly trans, 99%

$34.66 - $134.41

Chemical Identifiers

Chemical Name or Material 2,5-Dimethoxycinnamic acid
CAS 10538-51-9
Name Note predominantly trans
Molecular Formula C11H12O4
Formula Weight 208.21
MDL Number MFCD00004378
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2164906

  SDS
alfa aesar™
B2164906
5g Each for $34.66

AAB2164914

  SDS
alfa aesar™
B2164914
25g Each for $134.41
Description & Specifications

Specifications

Formula Weight 208.21
Chemical Name or Material 2,5-Dimethoxycinnamic acid
Name Note predominantly trans
Assay 99%
Molecular Formula C11H12O4
Melting Point 147° to 150°C
MDL Number MFCD00004378
CAS 10538-51-9
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.