missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethoxybenzylamine, 97%, Maybridge™

Chemical Identifiers

CAS 3275-95-4
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052811
InChI Key UWMCHDDHXMFKMA-UHFFFAOYSA-N
Synonym 1-2,5-dimethoxyphenyl methanamine, 2,5-dimethoxy-benzylamine, 2,5-dimethoxybenzenemethanamine, 2,5-dimethoxybenzylamine, 2,5-dimethoxyphenyl methanamine, acmc-1cs2n, benzenemethanamine, 2,5-dimethoxy, benzylamine, 2,5-dimethoxy, pubchem7396, zilain
PubChem CID 18652
IUPAC Name (2,5-dimethoxyphenyl)methanamine
SMILES COC1=CC(=C(C=C1)OC)CN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
TL00178DA
SDS View Documents
Fisher Scientific Company
TL00178DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

3275-95-4
167.208
UWMCHDDHXMFKMA-UHFFFAOYSA-N
18652
COC1=CC(=C(C=C1)OC)CN
C9H13NO2
MFCD00052811
1-2,5-dimethoxyphenyl methanamine, 2,5-dimethoxy-benzylamine, 2,5-dimethoxybenzenemethanamine, 2,5-dimethoxybenzylamine, 2,5-dimethoxyphenyl methanamine, acmc-1cs2n, benzenemethanamine, 2,5-dimethoxy, benzylamine, 2,5-dimethoxy, pubchem7396, zilain
(2,5-dimethoxyphenyl)methanamine

Specifications

1.1100g/mL
1g
97.0
C9H13NO2
MFCD00052811
UWMCHDDHXMFKMA-UHFFFAOYSA-N
(2,5-dimethoxyphenyl)methanamine
18652
97%
Amber Glass Bottle
3275-95-4
97%
NH2CH2C6H3(OCH3)2
1-2,5-dimethoxyphenyl methanamine, 2,5-dimethoxy-benzylamine, 2,5-dimethoxybenzenemethanamine, 2,5-dimethoxybenzylamine, 2,5-dimethoxyphenyl methanamine, acmc-1cs2n, benzenemethanamine, 2,5-dimethoxy, benzylamine, 2,5-dimethoxy, pubchem7396, zilain
COC1=CC(=C(C=C1)OC)CN
167.208
167.21
2, 5-Dimethoxybenzylamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS