missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethoxybenzylamine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company TL00178DA

 View more versions of this product

Catalog No. TL00178DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

167.21
97%
97.0
2, 5-Dimethoxybenzylamine
3275-95-4
MFCD00052811
1g
1.1100g/mL
C9H13NO2
NH2CH2C6H3(OCH3)2
97%
Amber Glass Bottle

Chemical Identifiers

3275-95-4
97%
NH2CH2C6H3(OCH3)2
MFCD00052811
97%
97.0
C9H13NO2
167.21
2, 5-Dimethoxybenzylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger