missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 182200250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 5-Dimethoxybenzyl alcohol | |
33524-31-1 | |
100.0 | |
122°C to 125°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5460 to 1.548 | |
MFCD00004615 | |
2,5-dimethoxybenzyl alcohol, 2,5-dimethoxyphenyl methanol, benzenemethanol, 2,5-dimethoxy, acmc-1ckmt, 2,5-dimethoxybenzylalcohol, 2,5-dimethoxylbenzylalcohol, benzenemethanol,2,5-dimethoxy, 2,5-dimethoxyphenyl-methanol, wgqmuabrzguaos-uhfffaoysa, 2,5-dimethoxyphenyl methanol # | |
COC1=CC(=C(C=C1)OC)CO | |
168.19 | |
168.19 | |
Viscous Liquid |
99% | |
98.5 | |
Colorless to Yellow | |
>110°C | |
98.5% min. (GC) | |
C9H12O3 | |
(CH3O)2C6H3CH2OH | |
25 mL | |
WGQMUABRZGUAOS-UHFFFAOYSA-N | |
(2,5-dimethoxyphenyl)methanol | |
118495 | |
99% |
Chemical Identifiers
33524-31-1 | |
168.19 | |
WGQMUABRZGUAOS-UHFFFAOYSA-N | |
118495 | |
COC1=CC(=C(C=C1)OC)CO |
C9H12O3 | |
MFCD00004615 | |
2,5-dimethoxybenzyl alcohol, 2,5-dimethoxyphenyl methanol, benzenemethanol, 2,5-dimethoxy, acmc-1ckmt, 2,5-dimethoxybenzylalcohol, 2,5-dimethoxylbenzylalcohol, benzenemethanol,2,5-dimethoxy, 2,5-dimethoxyphenyl-methanol, wgqmuabrzguaos-uhfffaoysa, 2,5-dimethoxyphenyl methanol # | |
(2,5-dimethoxyphenyl)methanol |
Safety and Handling
EINECSNumber : 251-562-0
TSCA : TSCA
RUO – Research Use Only