Please login to your online account to display your discounted pricing

2,5-Dimethoxybenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethoxybenzoic acid
Name Note 98%
CAS 2785-98-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10O4
Linear Formula (CH3O)2C6H3COOH
Formula Weight 182.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC339260010

  SDS
thermo fisher scientific
339260010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC339260050

  SDS
thermo fisher scientific
339260050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 182.18
Physical Form Fine Crystalline Powder
Quantity 1g
Color White
EINECS Number 220-503-0
TSCA TSCA
Melting Point 74° to 78°C
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
CAS Min % 97.5
Infrared Spectrum Authentic
Name Note 98%
Chemical Name or Material 2, 5-Dimethoxybenzoic acid
Linear Formula (CH3O)2C6H3COOH
CAS 2785-98-0
Assay Percent Range 98%
Molecular Formula C9H10O4
MDL Number MFCD00002436
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.