Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxybenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethoxybenzenesulfonyl chloride
CAS 1483-28-9
Molecular Formula C8H9ClO4S
Formula Weight 236.68
MDL Number MFCD00024872
UN Number UN3261
Beilstein 2648547
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1128806

  SDS
alfa aesar™
A1128806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1128814

  SDS
alfa aesar™
A1128814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1128822

  SDS
alfa aesar™
A1128822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.68
Chemical Name or Material 2,5-Dimethoxybenzenesulfonyl chloride
Assay 98%
Molecular Formula C8H9ClO4S
Melting Point 111° to 116°C
MDL Number MFCD00024872
CAS 1483-28-9
UN Number UN3261
Beilstein 2648547
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.