Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethoxybenzenesulfonamide, 97%

$152.39 - $591.08

Chemical Identifiers

Chemical Name or Material 2,5-Dimethoxybenzenesulfonamide
CAS 19116-90-6
Molecular Formula C8H11NO4S
Formula Weight 217.25
MDL Number MFCD00833415
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2264903

 
alfa aesar™
B2264903
1g Each for $152.39

AAB2264906

 
alfa aesar™
B2264906
5g Each for $591.08
Description & Specifications

Specifications

Formula Weight 217.25
Chemical Name or Material 2,5-Dimethoxybenzenesulfonamide
Assay 97%
Molecular Formula C8H11NO4S
Melting Point 145° to 149°C
MDL Number MFCD00833415
CAS 19116-90-6
Quantity 1g
RTECS Number DB2030000