Learn More
2,5-Dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1992814
Description
2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.
Solubility
Soluble in chloroform and methanol. Slightly soluble in water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
2,5-Dimethoxybenzaldehyde | |
47°C to 51°C | |
>110°C (230°F) | |
MFCD00003314 | |
509301 | |
benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde | |
AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
2,5-dimethoxybenzaldehyde | |
66726 | |
≥98% |
93-02-7 | |
146°C (10 mmHg) | |
C9H10O3 | |
25 g | |
Air Sensitive | |
Soluble in chloroform and methanol. Slightly soluble in water. | |
COC1=CC(=C(C=C1)OC)C=O | |
166.176 | |
166.18 |
Chemical Identifiers
93-02-7 | |
166.176 | |
AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
66726 | |
COC1=CC(=C(C=C1)OC)C=O |
C9H10O3 | |
MFCD00003314 | |
benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde | |
2,5-dimethoxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
H315-H319-H335
EINECSNumber : 202-211-5
RTECSNumber : CU5740500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only