missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethoxybenzaldehyde, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC115395000
Specifications
2, 5-Dimethoxybenzaldehyde | |
Authentic | |
0.5% max. | |
Yellow to Beige | |
500g | |
96.0 | |
96% min. (GC) | |
(CH3O)2C6H3CHO | |
2,5-dimethoxy benzaldehyde, 2,5-dimethoxybenzald, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde, 5-dimethoxy benzaldehyde, benzaldehyde, 2,5-dimethoxy, gentisic aldehyde, pubchem2176, unii-w49s1ppl78 | |
AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
2,5-dimethoxybenzaldehyde | |
66726 | |
Crystalline Powder, Crystals and/or Chunks |
113°C | |
Glass bottle | |
146.0°C (10.0 mmHg) | |
45.0°C to 50.0°C | |
93-02-7 | |
100.0 | |
C9H10O3 | |
08,245 | |
Solubility in water: 795mg/l (25°C). Other solubilities: soluble in most common organic solvents | |
COC1=CC(=C(C=C1)OC)C=O | |
166.18 | |
166.18 | |
97% |
Chemical Identifiers
93-02-7 | |
166.18 | |
2,5-dimethoxy benzaldehyde, 2,5-dimethoxybenzald, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde, 5-dimethoxy benzaldehyde, benzaldehyde, 2,5-dimethoxy, gentisic aldehyde, pubchem2176, unii-w49s1ppl78 | |
2,5-dimethoxybenzaldehyde |
C9H10O3 | |
AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
66726 | |
COC1=CC(=C(C=C1)OC)C=O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 202-211-5