missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Dimethoxyacetophenone, 99%

Manufacturer:  Alfa Aesar™ A1497322

 View more versions of this product

Catalog No. AAA1497322



Specifications

Specifications

2',5'-Dimethoxyacetophenone
Characteristic
18°C to 20°C
1201-38-3
MFCD00008728
2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
CC(=O)C1=C(C=CC(=C1)OC)OC
180.203
180.2
>110°C (230°F)
154°C to 156°C (10mmHg)
100g
C10H12O3
2047423
FAXUIYJKGGUCBO-UHFFFAOYSA-N
1-(2,5-dimethoxyphenyl)ethanone
70991
99%

Chemical Identifiers

1201-38-3
180.203
FAXUIYJKGGUCBO-UHFFFAOYSA-N
70991
CC(=O)C1=C(C=CC(=C1)OC)OC
C10H12O3
MFCD00008728
2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
1-(2,5-dimethoxyphenyl)ethanone
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents