Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Dimethoxyacetophenone, 99%

$37.41 - $321.72

Chemical Identifiers

Chemical Name or Material 2',5'-Dimethoxyacetophenone
CAS 1201-38-3
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00008728
Beilstein 2047423
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1497306

 
alfa aesar™
A1497306
5g Each for $37.41

AAA1497322

 
alfa aesar™
A1497322
100g Each for $321.72

AAA1497314

 
alfa aesar™
A1497314
25g Each for $115.45
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material 2',5'-Dimethoxyacetophenone
TSCA TSCA
Assay 99%
Molecular Formula C10H12O3
Melting Point 18° to 20°C
Boiling Point 154° to 156°C (10mmHg)
MDL Number MFCD00008728
CAS 1201-38-3
Beilstein 2047423
Quantity 5g
Flash Point >110°C (230°F)
Odor Characteristic