Please login to your online account to display your discounted pricing

2,5-Dimethoxy-4-nitroaniline, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  188670050

 View more versions of this product

Catalog No. AC188670050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 153° to 157°C
Color Dark khaki
Quantity 5g
Chemical Name or Material 2, 5-Dimethoxy-4-nitroaniline
CAS 6313-37-7
CAS Min % 98.0
CAS Max % 100.0
Physical Form Dark khaki granular crystalline powder
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C8H10N2O4
Linear Formula O2NC6H2(OCH3)2NH2
Formula Weight 198.18g/mol
MDL Number MFCD00007361
Beilstein 13, 789
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 5-Dimethoxy-4-nitroaniline
CAS 6313-37-7
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C8H10N2O4
Linear Formula O2NC6H2(OCH3)2NH2
Formula Weight 198.18g/mol
MDL Number MFCD00007361
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents