Please login to your online account to display your discounted pricing

2,5-Dimethoxy-4-nitroaniline, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethoxy-4-nitroaniline
CAS 6313-37-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H10N2O4
Linear Formula O2NC6H2(OCH3)2NH2
Formula Weight 198.18
MDL Number MFCD00007361
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188670050

  SDS
thermo fisher scientific
AC188670050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC188670010

  SDS
thermo fisher scientific
188670010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Granular Crystalline Powder
Assay ≥98.0%
Formula Weight 198.18
Quantity 5g
Color Dark khaki
Melting Point 153° to 157°C
Beilstein 13, 789
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 2, 5-Dimethoxy-4-nitroaniline
Packaging Glass bottle
Assay Percent Range 98+%
Molecular Formula C8H10N2O4
Linear Formula O2NC6H2(OCH3)2NH2
Infrared Spectrum Authentic
MDL Number MFCD00007361
CAS 6313-37-7
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.