missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethoxy-4-nitroaniline, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 188670010

 View more versions of this product

Catalog No. AC188670010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 5-Dimethoxy-4-nitroaniline
Glass bottle
153°C to 157°C
6313-37-7
100.0
C8H10N2O4
MFCD00007361
3,6-dimethoxy-4-nitroaniline, benzenamine, 2,5-dimethoxy-4-nitro, aniline, 2,5-dimethoxy-4-nitro, 2,5-dimethoxy-4-nitrophenylamine, pubchem16730, acmc-209nd8, aniline,5-dimethoxy-4-nitro, paragos 500036, 2,5-dimethoxy-4-nitro-aniline, attercop-chm at114495
COC1=CC(=C(C=C1N)OC)[N+](=O)[O-]
198.178
198.18
≥98.0%
Authentic
Khaki
1g
98.0
98+%
O2NC6H2(OCH3)2NH2
13, 789
ZTQGTYFYOODGOQ-UHFFFAOYSA-N
2,5-dimethoxy-4-nitroaniline
80581
Granular Crystalline Powder

Chemical Identifiers

6313-37-7
198.178
ZTQGTYFYOODGOQ-UHFFFAOYSA-N
80581
COC1=CC(=C(C=C1N)OC)[N+](=O)[O-]
C8H10N2O4
MFCD00007361
3,6-dimethoxy-4-nitroaniline, benzenamine, 2,5-dimethoxy-4-nitro, aniline, 2,5-dimethoxy-4-nitro, 2,5-dimethoxy-4-nitrophenylamine, pubchem16730, acmc-209nd8, aniline,5-dimethoxy-4-nitro, paragos 500036, 2,5-dimethoxy-4-nitro-aniline, attercop-chm at114495
2,5-dimethoxy-4-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents