missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethoxy-3-tetrahydrofurancarboxaldehyde, 90%, Tech., mixture of isomers, ACROS Organics™
Chemical Identifiers
CAS
50634-05-4
Molecular Formula
C7 H12 O4
Molecular Weight (g/mol)
160.17
MDL Number
MFCD00010283
InChI Key
QMIGEDXMDGEZSR-UHFFFAOYSA-N
Synonym
2,5-dimethoxy-3-tetrahydrofurancarbaldehyde, 2,5-dimethoxy-3-tetrahydrofurancarboxaldehyde, 2,5-dimethoxy-4-tetrahydrofurancarbaldehyde, 2,5-dimethoxy-tetrahydrofuran-3-carboxaldehyde, 2,5-dimethoxytetrahydrofuran-3-aldehyde, 2,5-dimethoxytetrahydrofuran-3-carbaldehyde, 3-formyl-2,5-dimethoxytetrahydrofuran, 3-furancarboxaldehyde,tetrahydro-2,5-dimethoxy, acmc-20aoau, tetrahydro-2,5-dimethoxyfuran-3-carbaldehyde
PubChem CID
567310
IUPAC Name
2,5-dimethoxyoxolane-3-carbaldehyde
SMILES
COC1CC(C(O1)OC)C=O
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
AC346470010
SDS
View Documents
Thermo Fisher Scientific
346470010
1g
Glass bottle
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Specifications
Chemical Identifiers
CAS
50634-05-4
Molecular Weight (g/mol)
160.17
InChI Key
QMIGEDXMDGEZSR-UHFFFAOYSA-N
PubChem CID
567310
SMILES
COC1CC(C(O1)OC)C=O
Molecular Formula
C7 H12 O4
MDL Number
MFCD00010283
Synonym
2,5-dimethoxy-3-tetrahydrofurancarbaldehyde, 2,5-dimethoxy-3-tetrahydrofurancarboxaldehyde, 2,5-dimethoxy-4-tetrahydrofurancarbaldehyde, 2,5-dimethoxy-tetrahydrofuran-3-carboxaldehyde, 2,5-dimethoxytetrahydrofuran-3-aldehyde, 2,5-dimethoxytetrahydrofuran-3-carbaldehyde, 3-formyl-2,5-dimethoxytetrahydrofuran, 3-furancarboxaldehyde,tetrahydro-2,5-dimethoxy, acmc-20aoau, tetrahydro-2,5-dimethoxyfuran-3-carbaldehyde
IUPAC Name
2,5-dimethoxyoxolane-3-carbaldehyde
Specifications
Density
1.1290g/mL
Infrared Spectrum
Authentic
Specific Gravity
1.129
Color
Yellow to Brown
CAS
50634-05-4
CAS Max %
100.0
Molecular Formula
C7 H12 O4
Synonym
2,5-dimethoxy-3-tetrahydrofurancarbaldehyde, 2,5-dimethoxy-3-tetrahydrofurancarboxaldehyde, 2,5-dimethoxy-4-tetrahydrofurancarbaldehyde, 2,5-dimethoxy-tetrahydrofuran-3-carboxaldehyde, 2,5-dimethoxytetrahydrofuran-3-aldehyde, 2,5-dimethoxytetrahydrofuran-3-carbaldehyde, 3-formyl-2,5-dimethoxytetrahydrofuran, 3-furancarboxaldehyde,tetrahydro-2,5-dimethoxy, acmc-20aoau, tetrahydro-2,5-dimethoxyfuran-3-carbaldehyde
SMILES
COC1CC(C(O1)OC)C=O
Molecular Weight (g/mol)
160.17
Formula Weight
160.17
Percent Purity
≥88%
Chemical Name or Material
2, 5-Dimethoxy-3-tetrahydrofurancarboxaldehyde, Mixture of isomers
Flash Point
81°C
Packaging
Glass bottle
Boiling Point
89°C (12.0mmHg)
Quantity
1g
CAS Min %
88.0
Assay Percent Range
90%
MDL Number
MFCD00010283
InChI Key
QMIGEDXMDGEZSR-UHFFFAOYSA-N
IUPAC Name
2,5-dimethoxyoxolane-3-carbaldehyde
PubChem CID
567310
Physical Form
Liquid
Grade
Technical
Safety and Handling
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS