Learn More
2,5-Dimercapto-1,3,4-thiadiazole, 97% (dry wt.), water <3%, Thermo Scientific Chemicals
$85.92 - $340.89
Chemical Identifiers
CAS | 1072-71-5 |
---|---|
Molecular Formula | C2H2N2S3 |
Molecular Weight (g/mol) | 150.23 |
MDL Number | MFCD00003103 |
InChI Key | BIGYLAKFCGVRAN-UHFFFAOYSA-N |
Synonym | 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i |
PubChem CID | 2723630 |
IUPAC Name | 1,3,4-thiadiazolidine-2,5-dithione |
SMILES | S=C1NNC(=S)S1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1261518
|
Thermo Scientific Chemicals
A1261518 |
50 g |
Each for $85.92
|
|
|||||
AAA1261530
|
Thermo Scientific Chemicals
A1261530 |
250 g |
Each for $340.89
|
|
|||||
Description
2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
1072-71-5 | |
150.23 | |
BIGYLAKFCGVRAN-UHFFFAOYSA-N | |
2723630 | |
S=C1NNC(=S)S1 |
C2H2N2S3 | |
MFCD00003103 | |
1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i | |
1,3,4-thiadiazolidine-2,5-dithione |
Specifications
1072-71-5 | |
C2H2N2S3 | |
50 g | |
1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i | |
BIGYLAKFCGVRAN-UHFFFAOYSA-N | |
1,3,4-thiadiazolidine-2,5-dithione | |
2723630 | |
98% |
∼162°C (decomposition) | |
MFCD00003103 | |
113444 | |
Soluble in water. | |
S=C1NNC(=S)S1 | |
150.23 | |
150.24 | |
2,5-Dimercapto-1,3,4-thiadiazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-014-1
RTECSNumber : XI3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only