Learn More
2,5-Diiodothiophene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1244306
Description
2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.
Solubility
Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
2,5-Diiodothiophene | |
39°C to 43°C | |
C4H2I2S | |
5 g | |
Light sensitive | |
Insoluble in water. | |
C1=C(SC(=C1)I)I | |
335.929 | |
335.93 |
625-88-7 | |
139°C to 140°C (15 mmHg) | |
MFCD00014525 | |
109915 | |
thiophene, 2,5-diiodo, 2,5-diiodthiophen, thiophene,5-diiodo, pubchem5529, maybridge1_002904, acmc-1b0lo, 2,5-diiodothiophene, bidd:gt0169, thiophene, 2,5-diiodo-8ci 9ci | |
PNYWRAHWEIOAGK-UHFFFAOYSA-N | |
2,5-diiodothiophene | |
69368 | |
99% |
Chemical Identifiers
625-88-7 | |
335.929 | |
PNYWRAHWEIOAGK-UHFFFAOYSA-N | |
69368 | |
C1=C(SC(=C1)I)I |
C4H2I2S | |
MFCD00014525 | |
thiophene, 2,5-diiodo, 2,5-diiodthiophen, thiophene,5-diiodo, pubchem5529, maybridge1_002904, acmc-1b0lo, 2,5-diiodothiophene, bidd:gt0169, thiophene, 2,5-diiodo-8ci 9ci | |
2,5-diiodothiophene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 210-915-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only