missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diiodothiophene, 99%

Manufacturer:  Alfa Aesar™ A1244314

 View more versions of this product

Catalog No. AAA1244314



Description

Description

Applications
2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.

Solubility
Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

2,5-Diiodothiophene
99%
139°C to 140°C (15mmHg)
25g
109915
C1=C(SC(=C1)I)I
335.929
335.93
625-88-7
39°C to 43°C
C4H2I2S
MFCD00014525
PNYWRAHWEIOAGK-UHFFFAOYSA-N
2,5-diiodothiophene
69368

Chemical Identifiers

625-88-7
335.929
PNYWRAHWEIOAGK-UHFFFAOYSA-N
2,5-diiodothiophene
C4H2I2S
MFCD00014525
69368
C1=C(SC(=C1)I)I
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2°C to 8°C

SDS Documents