Learn More
2,5-Dihydroxyterephthalic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6488814
Description
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in hot dimethylformamide.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2,5-Dihydroxyterephthalic acid | |
>300°C | |
C8H6O6 | |
25 g | |
1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid | |
OYFRNYNHAZOYNF-UHFFFAOYSA-N | |
2,5-dihydroxyterephthalic acid | |
69131 | |
97% |
610-92-4 | |
Gray to Green | |
MFCD00132933 | |
2696052 | |
Soluble in hot dimethylformamide. | |
C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O | |
198.13 | |
198.13 | |
Solid |
Chemical Identifiers
610-92-4 | |
198.13 | |
OYFRNYNHAZOYNF-UHFFFAOYSA-N | |
69131 | |
C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O |
C8H6O6 | |
MFCD00132933 | |
1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid | |
2,5-dihydroxyterephthalic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-239-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only