missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dihydroxyphenyl(diphenyl)phosphine Oxide 97.0+%, TCI America™
$106.17 - $343.41
Chemical Identifiers
CAS | 13291-46-8 |
---|---|
Molecular Formula | C18H15O3P |
Molecular Weight (g/mol) | 310.289 |
InChI Key | LLOXZCFOAUCDAE-UHFFFAOYSA-N |
PubChem CID | 2783495 |
IUPAC Name | 2-diphenylphosphorylbenzene-1,4-diol |
SMILES | C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=C(C=CC(=C3)O)O |
Chemical Identifiers
13291-46-8 | |
310.289 | |
2783495 | |
C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=C(C=CC(=C3)O)O |
C18H15O3P | |
LLOXZCFOAUCDAE-UHFFFAOYSA-N | |
2-diphenylphosphorylbenzene-1,4-diol |
Specifications
13291-46-8 | |
White-Yellow | |
5 g | |
LLOXZCFOAUCDAE-UHFFFAOYSA-N | |
2-diphenylphosphorylbenzene-1,4-diol | |
2783495 | |
≥97.0% (GC) | |
2,5-Dihydroxyphenyl(diphenyl)phosphine Oxide |
214°C | |
C18H15O3P | |
3077 | |
C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=C(C=CC(=C3)O)O | |
310.289 | |
310.29 | |
Crystalline Powder |
Safety and Handling
TSCA : No