missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dihydroxybenzaldehyde, 98+%

Manufacturer:  Alfa Aesar™ A1556514

 View more versions of this product

Catalog No. AAA1556514



Description

Description

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air.
Specifications

Specifications

2,5-Dihydroxybenzaldehyde
25g
C7H6O3
1363961
CLFRCXCBWIQVRN-UHFFFAOYSA-N
2,5-dihydroxybenzaldehyde
70949
138.12
97°C to 101°C
1194-98-5
MFCD00003333
gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy
C1=CC(=C(C=C1O)C=O)O
138.122
CHEBI:28508
≥98%

Chemical Identifiers

1194-98-5
138.122
CLFRCXCBWIQVRN-UHFFFAOYSA-N
70949
2,5-dihydroxybenzaldehyde
C7H6O3
MFCD00003333
gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy
CHEBI:28508
C1=CC(=C(C=C1O)C=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents