missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Dihydroxyacetophenone, 98+%

Manufacturer:  Alfa Aesar™ A1218506

 View more versions of this product

Catalog No. AAA1218506


 

Specifications

Specifications

2',5'-Dihydroxyacetophenone
5g
≥98%
MFCD00002343
2-Acetylhydroquinone
CC(=O)C1=C(C=CC(=C1)O)O
152.149
152.15
204°C to 208°C
490-78-8
C8H8O3
637903
WLDWSGZHNBANIO-UHFFFAOYSA-N
1-(2,5-dihydroxyphenyl)ethanone
10279

Chemical Identifiers

490-78-8
152.149
WLDWSGZHNBANIO-UHFFFAOYSA-N
10279
C8H8O3
MFCD00002343
2-Acetylhydroquinone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AM7700000

TSCA : TSCA

SDS Documents